missing translation for 'onlineSavingsMsg'
Learn More

Ethyl hexafluoroglutaryl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11328935
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328935 1 g 1g
11348935 250 mg 250mg
11338935 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11328935 Supplier Thermo Scientific Alfa Aesar Supplier No. L14049.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ethyl hexafluoroglutaryl chloride is used as a derivatising agent in gas chromatography. It is an active reagent used for the quantization of amphetamine and methamphetamine in urine by gas chromatography/mass spectroscopy (GC/MS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl hexafluoroglutaryl chloride is used as a derivatising agent in gas chromatography. It is an active reagent used for the quantization of amphetamine and methamphetamine in urine by gas chromatography/mass spectroscopy (GC/MS).

Solubility
Miscible with water.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, bases and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18381-53-8
Molecular Formula C7H5ClF6O3
Molecular Weight (g/mol) 286.55
MDL Number MFCD00054671
InChI Key OLRXGDHRDQKNGW-UHFFFAOYSA-N
PubChem CID 161251
IUPAC Name ethyl 5-chloro-2,2,3,3,4,4-hexafluoro-5-oxopentanoate
SMILES CCOC(=O)C(F)(F)C(F)(F)C(F)(F)C(Cl)=O

Specifications

Boiling Point 158°C to 160°C
Quantity 1 g
UN Number UN3265
Beilstein 1884844
Synonym 4-carbethoxyhexafluorobutyryl chloride, ethyl hexafluoroglutaryl chloride, pentanoic acid, 5-chloro-2,2,3,3,4,4-hexafluoro-5-oxo-, ethyl ester, 4ce-hfba, 4-carbethoxyhexafluorobutyryl, ethyl hexafluoroglutaryl chloride 250mg, 4-ethoxycarbonyl-4,4,3,3,2,2-hexafluorobutyryl chloride, pentanoic acid,5-chloro-2,2,3,3,4,4-hexafluoro-5-oxo-, ethyl ester
Formula Weight 286.56
Percent Purity 97%
Chemical Name or Material Ethyl hexafluoroglutaryl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.