missing translation for 'onlineSavingsMsg'
Learn More

Ethyl heptanoate, 98+%

Product Code. 11437766
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11437766 100 g 100g
11447766 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11437766 Supplier Thermo Scientific Alfa Aesar Supplier No. A14729.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 106-30-9 | C9H18O2 | 158.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-30-9
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00009538
InChI Key TVQGDYNRXLTQAP-UHFFFAOYSA-N
Synonym ethyl enanthate, oenanthic ether, wine oil, ethyl heptylate, grape oil, ethyl heptoate, heptanoic acid, ethyl ester, cognac oil, enanthylic ether, ethyl oenanthate
PubChem CID 7797
ChEBI CHEBI:86618
IUPAC Name ethyl heptanoate
SMILES CCCCCCC(=O)OCC

Specifications

Melting Point -66°C
Density 0.868
Boiling Point 188°C to 189°C
Flash Point 66°C (151°F)
Refractive Index 1.412
Quantity 100 g
Beilstein 1752311
Merck Index 14,3835
Formula Weight 158.24
Percent Purity ≥98%
Chemical Name or Material Ethyl heptanoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.