missing translation for 'onlineSavingsMsg'
Learn More

Ethyl heptafluorobutyrate, 98%

Product Code. 11462758
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462758 100 g 100g
11452758 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11462758 Supplier Thermo Scientific Alfa Aesar Supplier No. A16742.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 356-27-4 | C6H5F7O2 | 242.093 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 356-27-4
Molecular Formula C6H5F7O2
Molecular Weight (g/mol) 242.093
MDL Number MFCD00000434
InChI Key JVHJRIQPDBCRRE-UHFFFAOYSA-N
Synonym ethyl heptafluorobutyrate, ethyl perfluorobutyrate, heptafluorobutyric acid ethyl ester, butanoic acid, heptafluoro-, ethyl ester, ethyl heptafluorobutanoate, ethyl perfluorobutanoate, ethyl heptafluoro-n-butyrate, butyric acid, heptafluoro-, ethyl ester, butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, ethyl ester, pubchem12539
PubChem CID 9644
IUPAC Name ethyl 2,2,3,3,4,4,4-heptafluorobutanoate
SMILES CCOC(=O)C(C(C(F)(F)F)(F)F)(F)F

Specifications

Density 1.398
Boiling Point 95°C to 98°C
Flash Point 14°C (57°F)
Refractive Index 1.302
Quantity 100 g
UN Number UN3272
Beilstein 1794634
Formula Weight 242.09
Percent Purity 98%
Chemical Name or Material Ethyl heptafluorobutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.