missing translation for 'onlineSavingsMsg'
Learn More

Ethyl glycolate, 95%

Product Code. 11354218
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354218 10 g 10g
11344218 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11354218 Supplier Thermo Scientific Alfa Aesar Supplier No. 041475.09
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 623-50-7 | C4H8O3 | 104.11 g/mol

Ethyl glycolate, is an important building block for the preparation of agrochemicals and medicinal compounds. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff. It is also used in synthetic chemistry, discovery process and R&D. It also can be used as solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-50-7
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00021970
InChI Key ZANNOFHADGWOLI-UHFFFAOYSA-N
Synonym ethyl glycolate, ethyl hydroxyacetate, acetic acid, hydroxy-, ethyl ester, glycolic acid, ethyl ester, ethyl glycollate, hydroxyacetic acid ethyl ester, glycolic acid ethyl ester, unii-c306e91wfk, acetic acid, 2-hydroxy-, ethyl ester, 2-hydroxyacetic acid ethyl ester
PubChem CID 12184
IUPAC Name ethyl 2-hydroxyacetate
SMILES CCOC(=O)CO

Specifications

Color Colorless
Density 1.10 g/mL
Boiling Point 158°C to 159°C
Flash Point 61°C (143°F)
Linear Formula HOCH2CO2C2H5
Refractive Index 1.419
Quantity 10 g
Solubility Information It is very soluble in water,alcohols,acetone,acetates,slightly soluble in ethyl ether,sparingly soluble in hydrocarbon solvents,polymerization occurs above 50 C.
Formula Weight 104.11
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl glycolate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.