missing translation for 'onlineSavingsMsg'
Learn More

Ethyl glutaryl chloride, 97%

Product Code. 15445768
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15445768 25 mL 25mL
15435768 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15445768 Supplier Thermo Scientific Alfa Aesar Supplier No. H59443.AC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5205-39-0 | C7H11ClO3 | 178.612 g/mol

Ethyl glutaryl chloride is a main cationic monomer, it can bring quaternary ammonium salt

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl glutaryl chloride is a main cationic monomer, it can bring quaternary ammonium salt

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5205-39-0
Molecular Formula C7H11ClO3
Molecular Weight (g/mol) 178.612
MDL Number MFCD00013659
InChI Key KKJAQUGGQMCNJY-UHFFFAOYSA-N
Synonym ethyl 4-chloroformyl butyrate, ethyl 5-chloro-5-oxovalerate, glutaric acid monoethyl ester chloride, pentanoic acid, 5-chloro-5-oxo-, ethyl ester, ethyl 4-chlorocarbonyl butanoate, pubchem16647, ksc939c3b, gamma-carbethoxybutyryl chloride, glutaric acid monoethylester chloride, 5-chloro-5-oxovaleric acid ethyl ester
PubChem CID 78881
IUPAC Name ethyl 5-chloro-5-oxopentanoate
SMILES CCOC(=O)CCCC(=O)Cl

Specifications

Color Colorless
Density 1.139 g/mL
Boiling Point 68°C to 70°C (0.07 mmHg)
Linear Formula ClCO(CH2)3COOC2H5
Refractive Index 1.443
Quantity 25 mL
UN Number UN3265
Beilstein 1099817
Solubility Information Soluble in water.
Formula Weight 178.61
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl glutaryl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.