missing translation for 'onlineSavingsMsg'
Learn More

Ethyl gallate, MedChemExpress

Product Code. 16489096
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489096 100 mg 100mg
16402539 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16489096 Supplier Medchem Express Supplier No. HYN0525
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ethyl gallate is a nonflavonoid phenolic compound and also a scavenger of hydrogen peroxide.

Ethyl gallate is a nonflavonoid phenolic compound and also a scavenger of hydrogen peroxide.

Specifications

Chemical Name or Material Ethyl gallate
CAS 831-61-8
Color White
Purity Grade Notes Research
Molecular Formula C9H10O5
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (504.62 mM)
SMILES O=C(OCC)C1=CC(O)=C(O)C(O)=C1
Molecular Weight (g/mol) 198.17
Formula Weight 198.17
Percent Purity 98.94%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.