missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Formate, TRC
Spécification
Spécification
| Chemical Name or Material | Formic acid ethyl ester |
| CAS | 109-94-4 |
| Molecular Formula | C3 H6 O2 |
| Quantity | 5 g |
| Synonym | Formic Acid Ethyl Ester, Formic acid, ethyl ester, Areginal, Ethyl formate, Ethyl methanoate, Methanoic acid ethyl ester, NSC 406578, NSC 8828 |
| InChI Formula | InChI=1S/C3H6O2/c1-2-5-3-4/h3H,2H2,1H3 |
| SMILES | CCOC=O |
| IUPAC Name | ethyl formate |
| Molecular Weight (g/mol) | 74.08 |
| Formula Weight | 74.0368 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?