missing translation for 'onlineSavingsMsg'
Learn More

Ethyl formate, 98+%, pure

Product Code. 10245670 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal drum
Unit Size:
1 L
2.5 L
25 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10245670 1 L Glass bottle 1 L
10448680 500 mL Glass bottle 500mL
10478490 2.5 L Glass bottle 2.5 L
10544084 25 L Metal drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10245670 Supplier Thermo Scientific Acros Supplier No. 150670010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-94-4 | C3H6O2 | 74.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-94-4
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
MDL Number MFCD00003294
InChI Key WBJINCZRORDGAQ-UHFFFAOYSA-N
Synonym areginal, formic acid, ethyl ester, ethyl methanoate, formic acid ethyl ester, aethylformiat, ethyl formic ester, ethylformate, ethylformiaat, formic ether, ethylformiat
PubChem CID 8025
ChEBI CHEBI:52342
IUPAC Name ethyl formate
SMILES CCOC=O

Specifications

Melting Point -80.0°C
Density 0.9170g/mL
Boiling Point 54.0°C
Flash Point −20°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula HCO2C2H5
Refractive Index 1.3580 to 1.3610
Quantity 1 L
Beilstein 02, 19
Fieser 01,380; 02,197; 04,233; 12,226
Merck Index 15, 3862
Specific Gravity 0.917
Solubility Information Solubility in water: 110g/L (18°C). Other solubilities: freely soluble in ethylacetaat, isobutylacetaat,, butylacetaat, aceton, methylisobutylketon and, methylethylketon, miscible with benzene, ether and alcohol
Viscosity 0.4 mPa.s (20°C)
Formula Weight 74.08
Percent Purity 98+%
Grade Pure
Physical Form Liquid
Chemical Name or Material Ethyl formate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl formate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.