missing translation for 'onlineSavingsMsg'
Learn More

Ethyl formate, 97%

Product Code. 11409003
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409003 2500 mL 2500mL
11408940 100 mL 100mL
11498993 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409003 Supplier Thermo Scientific Alfa Aesar Supplier No. A11113.0F
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-94-4 | C3H6O2 | 74.08 g/mol

Ethyl formate is used as a solvent for cellulose nitrate, cellulose acetate, greases and oils. It is also used as a flavor for lemonade and essences. It is involved in the fumigation of dried fruits, as an alternative to methyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl formate is used as a solvent for cellulose nitrate, cellulose acetate, greases and oils. It is also used as a flavor for lemonade and essences. It is involved in the fumigation of dried fruits, as an alternative to methyl bromide.

Solubility
Miscible with water, benzene, alcohol, ether, acetone and most organic solvents.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-94-4
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
MDL Number MFCD00003294
InChI Key WBJINCZRORDGAQ-UHFFFAOYSA-N
Synonym areginal, formic acid, ethyl ester, ethyl methanoate, formic acid ethyl ester, aethylformiat, ethyl formic ester, ethylformate, ethylformiaat, formic ether, ethylformiat
PubChem CID 8025
ChEBI CHEBI:52342
IUPAC Name ethyl formate
SMILES CCOC=O

Specifications

Melting Point -81°C
Density 0.921
pH 4.1
Boiling Point 52°C to 54°C
Flash Point −19°C (−2°F)
Assay Percent Range 97%
Odor Fruit-like
Linear Formula HCO2CH2CH3
Refractive Index 1.36
Quantity 2500 mL
UN Number UN1190
Beilstein 906769
Sensitivity Moisture sensitive
Merck Index 14,3807
Solubility Information Miscible with water,benzene,alcohol,ether,acetone and most organic solvents.
Formula Weight 74.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl formate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.