Learn More
Ethyl fluoroacetate, 98%
Description
Ethyl fluoroacetate is used in the synthesis of 4-deoxy-4-fluoro-muscarines. It is also used as starting reagent for the synthesis of ethyl(diethoxyphosphoryl)fluoroacetate. and in development of an ammonolysis-based microencapsulation process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl fluoroacetate is used in the synthesis of 4-deoxy-4-fluoro-muscarines. It is also used as starting reagent for the synthesis of ethyl(diethoxyphosphoryl)fluoroacetate. and in development of an ammonolysis-based microencapsulation process.
Solubility
Decomposes in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
| Color | Colorless to Yellow |
| Density | 1.093 g/mL |
| Boiling Point | 119°C to 120°C |
| Linear Formula | FCH2CO2CH2CH3 |
| Refractive Index | 1.375 |
| Quantity | 10 g |
| UN Number | UN2929 |
| Beilstein | 1743761 |
| Merck Index | 14,4167 |
| Solubility Information | Decomposes in water. |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.