missing translation for 'onlineSavingsMsg'
Learn More

Ethyl ferulate, MedChemExpress

Product Code. 16478966
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478966 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16478966 Supplier Medchem Express Supplier No. HYN0061

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ethyl ferulate, a naturally lipophilic derivative of ferulic acid originally derived from giant fennel (F. communis), induces heme oxygenase-1 (HO-1) and protects rat neurons against oxidative stress. Ethyl ferulate also protects neurons against amyloid β peptide (1-42)-induced oxidative stress and neurotoxicity.

Ethyl ferulate, a naturally lipophilic derivative of ferulic acid originally derived from giant fennel (F. communis), induces heme oxygenase-1 (HO-1) and protects rat neurons against oxidative stress. Ethyl ferulate also protects neurons against amyloid β peptide (1-42)-induced oxidative stress and neurotoxicity.

Specifications

Chemical Name or Material Ethyl ferulate
CAS 4046-02-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C12H14O4
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (449.96 mM)
SMILES O=C(OCC)/C=C/C1=CC=C(O)C(OC)=C1
Molecular Weight (g/mol) 222.24
Formula Weight 222.24
Percent Purity 99.14%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.