missing translation for 'onlineSavingsMsg'
Learn More

Ethyl ethynyl ether, 50 wt% solution in hexanes

Product Code. 10105843 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10105843 25 g Glass bottle 25g
10037782 1 g Glass bottle 1g
10329120 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10105843 Supplier Thermo Scientific Acros Supplier No. 307390250
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 927-80-0 | C4H6O | 70.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 927-80-0 , 92112-69-1
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00009247
InChI Key WMYNMYVRWWCRPS-UHFFFAOYSA-N
Synonym ethoxyacetylene, ethoxyethyne, ethyl ethynyl ether, ethyne, ethoxy, ether, ethyl ethynyl, ethoxy-ethyne, ethyne, ethoxy-9ci, 1-ethoxyacetylene, ethoxyacetylene w/w solution in hexanes, ethoxy acetylene
PubChem CID 61239
IUPAC Name ethynoxyethane
SMILES CCOC#C

Specifications

Color Orange to Brown
Density 0.7180g/mL
Boiling Point 55.0°C to 56.0°C
Flash Point −28°C
Packaging Glass bottle
Linear Formula C2H5OC≡CH
Quantity 25 g
Specific Gravity 0.718
Formula Weight 70.09
Physical Form Liquid
Chemical Name or Material Ethyl ethynyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.