missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (ethoxymethylene)cyanoacetate, 98%, Thermo Scientific Chemicals

Catalog Number 10636882
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
94-05-3
C8H11NO3
169.18
MFCD00009136
KTMGNAIGXYODKQ-SREVYHEPSA-N
ethyl ethoxymethylene cyanoacetate, e-ethyl 2-cyano-3-ethoxyacrylate, ethyl 2-cyano-3-ethoxyacrylate, ethyl 3-ethoxy-2-cyanoacrylate, ethyl 2e-2-cyano-3-ethoxyprop-2-enoate, ethyl cyano ethoxymethylene acetate, 2-propenoic acid, 2-cyano-3-ethoxy-, ethyl ester, 2-cyano-3-ethoxy-2-propenoic acid ethyl ester, 2-cyano-3-ethoxyacrylic acid ethyl ester, ethyl 2e-2-cyano-3-ethoxy-2-propenoate
1715183
CCO\C=C(\C#N)C(=O)OCC
This item is not returnable. View return policy
94-05-3
C8H11NO3
169.18
MFCD00009136
KTMGNAIGXYODKQ-SREVYHEPSA-N
ethyl ethoxymethylene cyanoacetate, e-ethyl 2-cyano-3-ethoxyacrylate, ethyl 2-cyano-3-ethoxyacrylate, ethyl 3-ethoxy-2-cyanoacrylate, ethyl 2e-2-cyano-3-ethoxyprop-2-enoate, ethyl cyano ethoxymethylene acetate, 2-propenoic acid, 2-cyano-3-ethoxy-, ethyl ester, 2-cyano-3-ethoxy-2-propenoic acid ethyl ester, 2-cyano-3-ethoxyacrylic acid ethyl ester, ethyl 2e-2-cyano-3-ethoxy-2-propenoate
1715183
CCO\C=C(\C#N)C(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 48°C to 52°C
Color White to Yellow
Boiling Point 190°C to 191°C (30.0 mmHg)
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5OCH=C(CN)CO2C2H5
Quantity 500 g
Beilstein 03,47
Fieser 01,38
Solubility Information Solubility in water: <0.01g/L (20°C)
IUPAC Name ethyl (E)-2-cyano-3-ethoxyprop-2-enoate
Formula Weight 169.18
Percent Purity 98%
Physical Form Crystalline Mass or Crystals
Chemical Name or Material Ethyl (ethoxymethylene)cyanoacetate, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl (ethoxymethylene)cyanoacetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'