missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Ethanesulfonate, TRC

Product Code. 30372305
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372305 1 g 1g
30383137 2.5 g 2.5g
30372472 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30372305 Supplier Toronto Research Chemicals Supplier No. TRCE6765651G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1912-30-7
Molecular Formula C4H10O3S
Molecular Weight (g/mol) 138.19
Synonym Ethanesulfonic Acid Ethyl Ester, Ethylethane Sulfonate:
SMILES CCOS(CC)(=O)=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H10O3S/c1-3-7-8(5,6)4-2/h3-4H2,1-2H3
Formula Weight 138.0351
Chemical Name or Material Ethyl Ethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.