missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (E)-2-cyano-3-ethoxycrotonate, 98%

Product Code. 11352259
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352259 5 g 5g
11362259 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352259 Supplier Thermo Scientific Alfa Aesar Supplier No. H27244.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 932750-29-3 | C9H13NO3 | 183.207 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 932750-29-3
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.207
MDL Number MFCD01743485
InChI Key YKXFOGAYPIPTKF-BQYQJAHWSA-N
Synonym ethyl 2e-2-cyano-3-ethoxybut-2-enoate, ethyl e-2-cyano-3-ethoxycrotonate, ethyl 2-cyano-3-ethoxybut-2-enoate, 2-butenoic acid, 2-cyano-3-ethoxy-, ethyl ester, ethyl e-2-cyano-3-ethoxybut-2-enoate, e-ethyl 2-cyano-3-ethoxybut-2-enoate, ethyl e-2-cyano-3-ethoxy-2-butenoate, e-2-cyano-3-ethoxycrotonic acid ethyl ester, e-2-cyano-3-ethoxy-2-butenoic acid ethyl ester
PubChem CID 6387109
IUPAC Name ethyl (E)-2-cyano-3-ethoxybut-2-enoate
SMILES CCOC(=C(C#N)C(=O)OCC)C

Specifications

Melting Point 71°C to 73°C
Quantity 5 g
UN Number UN3439
Formula Weight 183.21
Percent Purity 98%
Chemical Name or Material Ethyl (E)-2-cyano-3-ethoxycrotonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.