missing translation for 'onlineSavingsMsg'
Learn More

Ethyl dodecanoate, 98%

Product Code. 11473484
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473484 100 mL 100mL
11483484 500 mL 500mL
11493484 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473484 Supplier Thermo Scientific Alfa Aesar Supplier No. A11188.AE
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 106-33-2 | C14H28O2 | 228.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-33-2
Molecular Formula C14H28O2
Molecular Weight (g/mol) 228.38
MDL Number MFCD00015065
InChI Key MMXKVMNBHPAILY-UHFFFAOYSA-N
Synonym ethyl laurate, dodecanoic acid, ethyl ester, ethyl laurinate, ethyl dodecylate, lauric acid ethyl ester, lauric acid, ethyl ester, ethyl laurate natural, dodecanoic acid ethyl ester, ethyl n-dodecanote, unii-f389d4md5k
PubChem CID 7800
ChEBI CHEBI:87427
IUPAC Name ethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCC

Specifications

Melting Point -2°C
Density 0.862
Boiling Point 268°C to 270°C
Flash Point 138°C (280°F)
Odor Fruit-like
Refractive Index 1.4315
Quantity 100 mL
Beilstein 1769671
Merck Index 14,3818
Formula Weight 228.38
Percent Purity 98%
Chemical Name or Material Ethyl dodecanoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.