missing translation for 'onlineSavingsMsg'
Learn More

Ethyl docosa-4,7,10,13,16,19-hexaenoate, MedChemExpress

Product Code. 16465447
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465447 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16465447 Supplier Medchem Express Supplier No. HYW011120
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ethyl cis-4,7,10,13,16,19-Docosahexaenoate, the ethyl ester of Docosahexaenoate (DHA), is enriched in the ethyl ester fraction by the selective alcoholysis of fatty acid ethyl esters originating from tuna oil with lauryl alcohol.

Ethyl cis-4,7,10,13,16,19-Docosahexaenoate, the ethyl ester of Docosahexaenoate (DHA), is enriched in the ethyl ester fraction by the selective alcoholysis of fatty acid ethyl esters originating from tuna oil with lauryl alcohol.

Specifications

Chemical Name or Material Ethyl docosa-4,7,10,13,16,19-hexaenoate
CAS 84494-72-4
Color Colorless
Purity Grade Notes Research
Molecular Formula C24H36O2
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (280.47 mM; Need ultrasonic)
SMILES CC/C=C/C/C=C/C/C=C/C/C=C/C/C=C/C/C=C/CCC(OCC)=O
Molecular Weight (g/mol) 356.55
Formula Weight 356.55
Percent Purity 97.0%
Grade Research
Physical Form Liquid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.