missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Ethyl diphenylphosphinite, 98%
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Description
Applications
Ethyl diphenylphosphinite is used in the synthesis of phosphoramides.
Solubility
Soluble in chloroform.
Notes
Air & Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Ethyl diphenylphosphinite is used in the synthesis of phosphoramides.
Solubility
Soluble in chloroform.
Notes
Air & Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Density | 1.064 |
| Boiling Point | 179°C to 180°C (14 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.59 |
| Quantity | 25 g |
| Beilstein | 1641750 |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Soluble in chloroform. |
| Formula Weight | 230.25 |
| Percent Purity | 98% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction