missing translation for 'onlineSavingsMsg'
Learn More

Ethyl dimethylpyruvate, 97%

Product Code. 15450198
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450198 25 g 25g
15440198 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15450198 Supplier Thermo Scientific Alfa Aesar Supplier No. H53413.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20201-24-5 | C7H12O3 | 144.17 g/mol

Ethyl dimethylpyruvate is used as an allyl halide reactant in the presence of indium to release hydroxy unsaturated carbonyl compounds. It was also used in the synthesis of (2SR,4SR)-5(S)-(N-Boc)-amino-6-cyclohexyl-4-hydoxy-2-isopropyl hexanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl dimethylpyruvate is used as an allyl halide reactant in the presence of indium to release hydroxy unsaturated carbonyl compounds. It was also used in the synthesis of (2SR,4SR)-5(S)-(N-Boc)-amino-6-cyclohexyl-4-hydoxy-2-isopropyl hexanoic acid.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20201-24-5
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00009122
InChI Key CKTYYUQUWFEUCO-UHFFFAOYSA-N
Synonym ethyl 3-methyl-2-oxobutyrate, ethyl dimethylpyruvate, ethyl 2-oxo-3-methylbutanoate, butanoic acid, 3-methyl-2-oxo-, ethyl ester, acmc-20akzg, ketovaline ethyl ester, pubchem18179, ethyl3-methyl-2-oxobutanoate, ethyl 2-methyl-2-oxobutyrate, ethyl-3-methyl-2-oxobutanoate
PubChem CID 88406
IUPAC Name ethyl 3-methyl-2-oxobutanoate
SMILES CCOC(=O)C(=O)C(C)C

Specifications

Density 0.989
Boiling Point 62°C (11 mmHg)
Flash Point 43°C (109°F)
Refractive Index 1.41
Quantity 25 g
UN Number UN3272
Beilstein 1756668
Solubility Information Slightly soluble in water.
Formula Weight 144.17
Percent Purity 97%
Chemical Name or Material Ethyl dimethylpyruvate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.