missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Digol, Pure, Fisher Chemical

Product Code. 10366420 Shop All Fisher Chemical Products
Click to view available options
Quantity:
2.5 L
Packaging:
Amber glass bottle
Unit Size:
2.5 L
This item is not returnable. View return policy

Product Code. 10366420

Brand: Fisher Chemical E/1300/17

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 111-90-0
Melting Point -80°C
Color Colorless
Boiling Point 197°C
Packaging Amber glass bottle
Molecular Formula C6H14O3
MDL Number 2872
Quantity 2.5 L
Synonym diethylene glycol monoethyl ether, 2-2-ethoxyethoxy ethanol, carbitol, transcutol, ethoxy diglycol, dioxitol, ethyl carbitol, ethyl digol, carbitol solvent, ethanol, 2-2-ethoxyethoxy
InChI Key XXJWXESWEXIICW-UHFFFAOYSA-N
SMILES CCOCCOCCO
IUPAC Name 2-(2-ethoxyethoxy)ethanol
Molecular Weight (g/mol) 134.175
PubChem CID 8146
ChEBI CHEBI:40572
Formula Weight 134.17g/mol
Physical Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.