missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl difluoroacetate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Ethyl difluoroacetate is used in pharmaceutical intermediates, acrylization oxidation and halogenating reaction of catalyst. Also ethyl difluoroacetate is used in preparing pyrrazolo-pyrimidine derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl difluoroacetate is used in pharmaceutical intermediates, acrylization oxidation and halogenating reaction of catalyst. Also ethyl difluoroacetate is used in preparing pyrrazolo-pyrimidine derivatives.
Solubility
Slightly soluble in water
Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Ethyl difluoroacetate is used in pharmaceutical intermediates, acrylization oxidation and halogenating reaction of catalyst. Also ethyl difluoroacetate is used in preparing pyrrazolo-pyrimidine derivatives.
Solubility
Slightly soluble in water
Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Density | 1.242 |
Boiling Point | 98°C to 99°C |
Flash Point | 25°C (77°F) |
Refractive Index | 1.3475 |
Quantity | 100 g |
UN Number | UN2924 |
Beilstein | 1748597 |
Solubility Information | Slightly soluble in water |
Formula Weight | 124.09 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'