missing translation for 'onlineSavingsMsg'
Learn More

Ethyl difluoroacetate, 98%

Product Code. 11453534
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453534 25 g 25g
11443534 5 g 5g
11463534 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453534 Supplier Thermo Scientific Alfa Aesar Supplier No. A11215.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 454-31-9 | C4H6F2O2 | 124.087 g/mol

Ethyl difluoroacetate is used in pharmaceutical intermediates, acrylization oxidation and halogenating reaction of catalyst. Also ethyl difluoroacetate is used in preparing pyrrazolo-pyrimidine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl difluoroacetate is used in pharmaceutical intermediates, acrylization oxidation and halogenating reaction of catalyst. Also ethyl difluoroacetate is used in preparing pyrrazolo-pyrimidine derivatives.

Solubility
Slightly soluble in water

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 454-31-9
Molecular Formula C4H6F2O2
Molecular Weight (g/mol) 124.087
MDL Number MFCD00013578
InChI Key GZKHDVAKKLTJPO-UHFFFAOYSA-N
Synonym ethyl difluoroacetate, acetic acid, difluoro-, ethyl ester, difluoroacetic acid ethyl ester, ethyldifluoroacetate, ethyl difluoroacetic, ethyl difluoro acetate, difluoro-acetic acid ethyl ester, ethyl difluoroacetic acid, acetic acid, 2,2-difluoro-, ethyl ester, ethyldifluoracetat
PubChem CID 9961
IUPAC Name ethyl 2,2-difluoroacetate
SMILES CCOC(=O)C(F)F

Specifications

Density 1.242
Boiling Point 98°C to 99°C
Flash Point 25°C (77°F)
Refractive Index 1.3475
Quantity 25 g
UN Number UN2924
Beilstein 1748597
Solubility Information Slightly soluble in water
Formula Weight 124.09
Percent Purity 98%
Chemical Name or Material Ethyl difluoroacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.