missing translation for 'onlineSavingsMsg'
Learn More

Ethyl difluoroacetate, 97%

Product Code. 10588273 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10588273 5 g Glass bottle 5g
10492673 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10588273 Supplier Thermo Scientific Acros Supplier No. 380390050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 454-31-9 | C4H6F2O2 | 124.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 454-31-9
Molecular Formula C4H6F2O2
Molecular Weight (g/mol) 124.09
MDL Number MFCD00013578
InChI Key GZKHDVAKKLTJPO-UHFFFAOYSA-N
Synonym ethyl difluoroacetate, acetic acid, difluoro-, ethyl ester, difluoroacetic acid ethyl ester, ethyldifluoroacetate, ethyl difluoroacetic, ethyl difluoro acetate, difluoro-acetic acid ethyl ester, ethyl difluoroacetic acid, acetic acid, 2,2-difluoro-, ethyl ester, ethyldifluoracetat
PubChem CID 9961
IUPAC Name ethyl 2,2-difluoroacetate
SMILES CCOC(=O)C(F)F

Specifications

Color Colorless
Density 1.2420g/mL
Boiling Point 97.0°C
Flash Point 25°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula F2CHCO2CH2CH3
Refractive Index 1.3465 to 1.3485
Quantity 5 g
UN Number 2924
Specific Gravity 1.242
Formula Weight 124.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl difluoroacetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl difluoroacetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.