missing translation for 'onlineSavingsMsg'
Learn More

Ethyl dichlorophosphate, 97%, Thermo Scientific Chemicals

Product Code. 10503314 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10503314 5 g Glass bottle 5g
10477490 25 g Glass bottle 25g
10744041 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10503314 Supplier Thermo Scientific Acros Supplier No. 118380050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1498-51-7
Molecular Formula C2H5Cl2O2P
Molecular Weight (g/mol) 162.94
MDL Number MFCD00002069
InChI Key YZBOZNXACBQJHI-UHFFFAOYSA-N
Synonym ethyl dichlorophosphate, ethyl phosphorodichloridate, phosphorodichloridic acid, ethyl ester, ethyldichlorophosphate, dichloroethoxyphosphine oxide, ethylphosphoric acid dichloride, dichlorophosphoric acid, ethyl ester, unii-4ip5dtw75u, ethylesterdichlorid german, hsdb 417
PubChem CID 15158
IUPAC Name 1-dichlorophosphoryloxyethane
SMILES CCOP(=O)(Cl)Cl

Specifications

Color Colorless to Brown
Density 1.3730g/mL
Boiling Point 167°C (758.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C2H5OP(O)Cl2
Refractive Index 1.4330 to 1.435
Quantity 5 g
Beilstein 01,332
Specific Gravity 1.373
Formula Weight 162.94
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl dichlorophosphate, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl dichlorophosphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.