missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Deoxycholate, TRC

Product Code. 30388253
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388253 100 mg
30376080 1 g
30377820 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388253 Supplier Toronto Research Chemicals Supplier No. TRCE915025100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69519-35-3
Molecular Formula C26H44O4
Molecular Weight (g/mol) 420.63
Synonym Deoxycholic Acid Ethyl Ester, (3α,5β,12α)-3,12-Dihydroxycholan-24-oic Acid Ethyl Ester
SMILES O[C@@H]1CC[C@@]2(C)[C@@](CC[C@]3([H])[C@]2([H])C[C@H](O)[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@@H](CCC(OCC)=O)C)([H])C1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C26H44O4/c1-5-30-24(29)11-6-16(2)20-9-10-21-19-8-7-17-14-18(27)12-13-25(17,3)22(19)15-23(28)26(20,21)4/h16-23,27-28H,5-15H2,1-4H3/t16-,17-,18-,19+,20-,21+,22+,23+,25+,26-/m1/s1
Formula Weight 420.324
Percent Purity >95%
Chemical Name or Material Ethyl Deoxycholate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.