missing translation for 'onlineSavingsMsg'
Learn More

Ethyl decanoate, 99%

Product Code. 11473404
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11473404

Brand: Thermo Scientific Alfa Aesar A10657.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-38-3 | C12H24O2 | 200.322 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-38-3
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.322
MDL Number MFCD00009581
InChI Key RGXWDWUGBIJHDO-UHFFFAOYSA-N
Synonym ethyl caprate, ethyl caprinate, ethyl decylate, decanoic acid ethyl ester, decanoic acid, ethyl ester, capric acid ethyl ester, ethyl n-decanoate, capric acid, ethyl ester, natural, unii-gy39fb86uo
PubChem CID 8048
ChEBI CHEBI:87430
IUPAC Name ethyl decanoate
SMILES CCCCCCCCCC(=O)OCC

Specifications

Melting Point -20°C
Density 0.862
Boiling Point 250°C to 252°C
Flash Point 102°C (216°F)
Refractive Index 1.425
Quantity 100 g
Beilstein 1762128
Merck Index 14,3776
Formula Weight 200.32
Percent Purity 99%
Chemical Name or Material Ethyl decanoate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.