missing translation for 'onlineSavingsMsg'
Learn More

(Ethyl-d5)triphenylphosphonium Bromide, TRC

Product Code. 30382869
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382869 25 mg 25mg
30384593 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30382869 Supplier Toronto Research Chemicals Supplier No. TRCE93195225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 875477-12-6
Molecular Formula C20H15D5BrP
Molecular Weight (g/mol) 376.28
Synonym Ethyl-d5-triphenylphosphonium Bromide, Triphenylethyl-d5-phosphonium Bromide, TEP-d5
IUPAC Name (ethyl-d5)triphenylphosphonium bromide
SMILES [2H]C([2H])([2H])C([2H])([2H])[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C20H20P.BrH/c1-2-21(18-12-6-3-7-13-18,19-14-8-4-9-15-19)20-16-10-5-11-17-20;/h3-17H,2H2,1H3;1H/q+1;/p-1/i1D3,2D2;
Formula Weight 375.08
Chemical Name or Material (Ethyl-d5)triphenylphosphonium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.