missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl-d5 Methanesulfonate, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Ethyl-D5 Methanesulfonate |
| CAS | 1219795-44-4 |
| Molecular Formula | C3 D5 H3 O3 S |
| Quantity | 0.25 g |
| Synonym | Ethan-1,1,2,2,2-d5-ol, 1-methanesulfonate (ACI), Pentadeuterioethyl methanesulfonate, Ethan-1,1,2,2,2-d5-ol 1-methanesulfonate, Ethyl-d5 Methanesulfonate, Ethyl Methanesulfonate-d5, Ethyl-D5 Methanesulfonate, Ethyl-d5 mesylate |
| InChI Formula | InChI=1 S/C3H8O3S/c1-3-6-7(2,4)5/h3H2,1-2H3/i1D3,3D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])OS(=O)(=O)C |
| IUPAC Name | 1,1,2,2,2-pentadeuterioethyl methanesulfonate |
| Molecular Weight (g/mol) | 129.1896 |
| Formula Weight | 129.0508 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction