missing translation for 'onlineSavingsMsg'
Learn More

Ethyl cyclopropanecarboxylate, 98%

Product Code. p-7027833
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416676 25 g 25g
11426676 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416676 Supplier Thermo Scientific Alfa Aesar Supplier No. B24870.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4606-07-9 | C6H10O2 | 114.14 g/mol

Ethyl cyclopropanecarboxylate is a reagent used in the addition of cyclopropane. It acts as annulation reagent for the conversion of aromatics to 2-methylindanones under Friedel-Crafts conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl cyclopropanecarboxylate is a reagent used in the addition of cyclopropane. It acts as annulation reagent for the conversion of aromatics to 2-methylindanones under Friedel-Crafts conditions.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4606-07-9
MDL Number MFCD00001282

Specifications

Density 0.96
Boiling Point 133°C to 134°C
Flash Point 18°C (64°F)
Molecular Formula C6H10O2
Refractive Index 1.42
Quantity 25 g
UN Number UN3272
Beilstein 2039752
Synonym cyclopropanecarboxylic acid ethyl ester, ethyl cyclopropylcarboxylate, ethyl cyclopropane carboxylate, cyclopropanecarboxylic acid, ethyl ester, pubchem11840, ethyl cylopropylcarboxylate, ethoxycarbonyl cyclopropane, ethylcyclopropane carboxylate, ksc497s5p, lddosdvzpsglfz-uhfffaoysa
Solubility Information Not miscible in water.
InChI Key LDDOSDVZPSGLFZ-UHFFFAOYSA-N
SMILES CCOC(=O)C1CC1
IUPAC Name ethyl cyclopropanecarboxylate
Molecular Weight (g/mol) 114.14
PubChem CID 78351
Formula Weight 114.15
Percent Purity 98%
Chemical Name or Material Ethyl cyclopropanecarboxylate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl cyclopropanecarboxylate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: Ethyl cyclopropanecarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.