missing translation for 'onlineSavingsMsg'
Learn More

Ethyl cyclohexanecarboxylate, 98+%

Product Code. 11357765
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357765 25 g 25g
11347765 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357765 Supplier Thermo Scientific Alfa Aesar Supplier No. L02622.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3289-28-9 | C9H16O2 | 156.225 g/mol

Ethyl cyclohexanecarboxylate is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl cyclohexanecarboxylate is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3289-28-9
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00041734
InChI Key JJOYCHKVKWDMEA-UHFFFAOYSA-N
Synonym cyclohexanecarboxylic acid, ethyl ester, ethyl cyclohexylformate, ethyl hexahydrobenzoate, cyclohexanecarboxylic acid ethyl ester, ethoxycarbonylcyclohexane, ethyl cyclohexylcarboxylate, ethyl cyclohexanoate, fema no. 3544, ethyl cyclohexylmethanoate, ethyl cyclohexanocarboxylate
PubChem CID 18686
IUPAC Name ethyl cyclohexanecarboxylate
SMILES CCOC(=O)C1CCCCC1

Specifications

Density 0.936
Boiling Point 194°C to 196°C
Refractive Index 1.442
Quantity 25 g
Beilstein 2042698
Solubility Information Insoluble in water.
Formula Weight 156.23
Percent Purity ≥98%
Chemical Name or Material Ethyl cyclohexanecarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.