missing translation for 'onlineSavingsMsg'
Learn More

Ethyl cyanoacetate, 98+%

Product Code. 11410413
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410413 500 g 500g
11420413 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410413 Supplier Thermo Scientific Alfa Aesar Supplier No. A11498.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 105-56-6 | C5H7NO2 | 113.12 g/mol

Cyanoacetate is a building block for the synthesis of heterocycles and are are used as drugs. It is also used as Pharmaceutical intermediates, or Oil soluble couplers of film raw material . It is also used as a reagent in labelled pyrimidine and purine synthesis. It is used as an intermediate of fine chemicals such as medicine and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyanoacetate is a building block for the synthesis of heterocycles and are are used as drugs. It is also used as Pharmaceutical intermediates, or Oil soluble couplers of film raw material . It is also used as a reagent in labelled pyrimidine and purine synthesis. It is used as an intermediate of fine chemicals such as medicine and dyestuff.

Solubility
It is slightly soluble in water. Mostly soluble in organic solvents such as chloroform, ethyl acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials strong acids, strong bases, strong oxidizing agents, strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-56-6
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
MDL Number MFCD00001940
InChI Key ZIUSEGSNTOUIPT-UHFFFAOYSA-N
Synonym ethyl cyanoacetate, ethyl cyanacetate, cyanoacetic acid ethyl ester, acetic acid, cyano-, ethyl ester, ethyl cyanoethanoate, cyanoacetic ester, cyanacetate ethyle, usaf kf-25, estere cianoacetico, ethyl cyano acetate
PubChem CID 7764
IUPAC Name ethyl 2-cyanoacetate
SMILES CCOC(=O)CC#N

Specifications

Melting Point -23°C
Density 1.063
Boiling Point 208°C to 210°C
Flash Point 109°C (228°F)
Odor Weak
Linear Formula NCCH2CO2CH2CH3
Refractive Index 1.418
Quantity 500 g
Beilstein 605871
Merck Index 14,3786
Solubility Information It is slightly soluble in water. Mostly soluble in organic solvents such as chloroform,ethyl acetate.
Formula Weight 113.12
Percent Purity ≥98%
Chemical Name or Material Ethyl cyanoacetate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl cyanoacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Ethyl cyanoacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.