missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Cyanoacetate-13C2, TRC

Product Code. 30380850
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380850 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380850 Supplier Toronto Research Chemicals Supplier No. TRCE67846225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Cyanoacetate-13C2
CAS 93664-96-1
Molecular Formula C313C2H7NO2
Quantity 25 mg
Synonym 2-Cyano-acetic Acid Ethyl Ester-13C2, (Ethoxycarbonyl)acetonitrile-13C2, Cyanoacetic alphacid epsilonthyl epsilonster-13C2, Cyanoacetic Ester-13C2, Ethyl 2-Cyanoacetate-13C2, Ethyl Cyanacetate-13C2, Ethyl Cyanoacetate-13C2, Ethyl Cyanoethanoate-13C2, Ethyl alpha-Cyanoacetate-13C2, Malonic Acid Ethyl Ester Nitrile-13C2, NSC 8844-13C2
InChI Formula InChI=1S/C5H7NO2/c1-2-8-5(7)3-4-6/h2-3H2,1H3/i4+1,5+1
SMILES CCO[13C](=O)C[13C]#N
IUPAC Name ethyl 2-(azanylidyne(1^{13}C)methyl)acetate
Molecular Weight (g/mol) 115.1
Formula Weight 115.0544
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.