missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Cyano(ethoxymethylene)acetate-13C3, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl Cyano(ethoxymethylene)acetate-13C3 |
| Molecular Formula | 13C3 C5 H11 N O3 |
| Quantity | 1 mg |
| Synonym | Ethyl Cyano(ethoxymethylidene)acetate-13C3, Ethyl alpha-(Ethoxymethylene)-alpha-cyanoacetate-13C3, NSC 27797-13C3, NSC 31579-13C3, NSC 62026-13C3, 2-(Ethoxymethylene)-2-cyanoacetic Acid Ethyl Ester-13C3, 2-Cyano-3-ethoxy-2-propenoic Acid Ethyl Ester-13C3, 2-Cyano-3-ethoxyacrylic Acid Ethyl Ester-13C3, Ethyl (Ethoxymethylene)cyanoacetate-13C3, Ethyl 2-(Ethoxymethylene)-2-cyanoacetate-13C3, Ethyl 2-Cyano-2-(ethoxymethylene)acetate-13C3, Ethyl 2-Cyano-3-(ethyloxy)-2-propenoate-13C3, Ethyl 2-Cyano-3-ethoxy-2-propenoate-13C3, Ethyl 2-Cyano-3-ethoxyacrylate-13C3, Ethyl 2-Cyano-3-ethoxypropenoate-13C3, Ethyl 2-Ethoxymethylidenecyanoacetate-13C3, Ethyl 3-Ethoxy-2-cyanoacrylate-13C3, 2-Cyano-3-ethoxyacrylic Acid Ethyl Ester-13C3 |
| InChI Formula | InChI=1S/C8H11NO3/c1-3-11-6-7(5-9)8(10)12-4-2/h6H,3-4H2,1-2H3/b7-6-/i5+1,6+1,8+1 |
| SMILES | CCO\[13CH]=C(\[13C]#N)/[13C](=O)OCC |
| IUPAC Name | ethyl (Z)-2-(azanylidyne(113C)methyl)-3-ethoxy(1,3-13C2)prop-2-enoate |
| Molecular Weight (g/mol) | 172.156 |
| Formula Weight | 172.084 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?