missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Crotonate, TRC

Product Code. 30379952
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30379952 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30379952 Supplier Toronto Research Chemicals Supplier No. TRCE67843510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Crotonate
CAS 623-70-1
Molecular Formula C6H10O2
Quantity 10 g
Synonym (E)-Crotonic Acid Ethyl Ester, (E)-2-Butenoic Acid Ethyl Ester, (E)-Ethyl but-2-enoate, Ethyl (E)-2-Butenoate, Ethyl (E)-Crotonate, Ethyl trans-2-Butenoate, Ethyl trans-Crotonate, trans-2-Butenoic Acid Ethyl Ester
InChI Formula InChI=1S/C6H10O2/c1-3-5-6(7)8-4-2/h3,5H,4H2,1-2H3/b5-3+
SMILES CCOC(=O)\C=C\C
IUPAC Name ethyl (E)-but-2-enoate
Molecular Weight (g/mol) 114.14
Formula Weight 114.0681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.