missing translation for 'onlineSavingsMsg'
Learn More

Ethyl cis-2-amino-1-cyclohexanecarboxylate hydrochloride, 95%

Product Code. 10791711 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
2.5 g
Packaging:
Glass Bottle
Unit Size:
1g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10791711 2.5 g Glass Bottle 2.5g
10781711 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10791711 Supplier Thermo Scientific Acros Supplier No. 265640025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1127-99-7 | C9H17NO2·HCl | 207.7 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1127-99-7
Molecular Formula C9H17NO2·HCl
Molecular Weight (g/mol) 207.7
MDL Number MFCD00144280
InChI Key XMQSOBPCWYVZSW-WLYNEOFISA-N
Synonym ethyl cis-2-amino-1-cyclohexanecarboxylate hydrochloride, cis-2-amino-cyclohexanecarboxylic acid ethyl ester hydrochloride, ethyl 1r,2s-2-aminocyclohexane-1-carboxylate hydrochloride, cis-ethyl 2-aminocyclohexanecarboxylate hydrochloride, 1r,2s-ethyl 2-aminocyclohexanecarboxylate hydrochloride, cyclohexanecarboxylic acid, 2-amino-, ethyl ester, hydrochloride, cis, ethyl cis-2-amino-1-cyclohexanecarboxylate hcl salt, ethyl cis-2-aminocyclohexanecarboxylate hydrochloride, ethyl 2s,1r-2-aminocyclohexanecarboxylate, chloride
PubChem CID 2724641
IUPAC Name ethyl (1R,2S)-2-aminocyclohexane-1-carboxylate;hydrochloride
SMILES CCOC(=O)C1CCCCC1N.Cl

Specifications

Melting Point 128°C to 135°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in methanol
Quantity 2.5 g
Formula Weight 207.7
Percent Purity 95%
Physical Form Crystalline Powder or Chunks
Chemical Name or Material Ethyl cis-2-amino-1-cyclohexanecarboxylate hydrochloride, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.