missing translation for 'onlineSavingsMsg'
Learn More

Ethyl cinnamate, 98+%

Product Code. 11459713
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459713 500 g 500g
11445363 100 g 100g
11469713 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459713 Supplier Thermo Scientific Alfa Aesar Supplier No. A12906.36
  • ONLINE EXCLUSIVE PRICE
    £67.30
    £60.65 / 500g
    Save £6.65
    9% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 103-36-6 | C11H12O2 | 176.215 g/mol

Ethyl cinnamate is used in medical intermediate, as a pharmaceutical intermediate and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl cinnamate is used in medical intermediate, as a pharmaceutical intermediate and in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, bases and reducing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 103-36-6
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00009189
InChI Key KBEBGUQPQBELIU-CMDGGOBGSA-N
Synonym ethyl cinnamate, ethyl 3-phenylacrylate, ethylcinnamate, cinnamic acid ethyl ester, ethylcinnamoate, cinnamic acid, ethyl ester, ethyl trans-cinnamate, e-ethyl cinnamate, ethyl 3-phenyl-2-propenoate, ethyl benzylideneacetate
PubChem CID 637758
ChEBI CHEBI:4895
IUPAC Name ethyl (E)-3-phenylprop-2-enoate
SMILES CCOC(=O)C=CC1=CC=CC=C1

Tekniske data

Melting Point 7°C
Density 1.049
Boiling Point 270°C to 272°C
Flash Point 135°C (275°F)
Assay Percent Range ≥98%
Refractive Index 1.559
Quantity 500 g
Beilstein 1238804
Merck Index 14,2299
Solubility Information Insoluble in water.
Formula Weight 176.22
Percent Purity ≥98%
Chemical Name or Material Ethyl Cinnamate
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • Ethyl cinnamate
Signalord
  • Warning
Farekategori
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Faresætning
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Sikkerhedssætning
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplerende oplysninger
  • MIXTURE LIST-Contain: Ethyl cinnamate

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.