missing translation for 'onlineSavingsMsg'
Learn More

Ethyl chrysanthemate, 95%, mixture of cis and trans

Product Code. 10127303 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10127303 25 g Glass bottle 25g
10437560 100 g Glass bottle 100g
10447560 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10127303 Supplier Thermo Scientific Acros Supplier No. 156060250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 97-41-6 | C12H20O2 | 196.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-41-6
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00001304
InChI Key VIMXTGUGWLAOFZ-UHFFFAOYSA-N
Synonym ethyl chrysanthemate, ethyl chrysanthemumate, chrysanthemic acid ethyl ester, cyclopropanecarboxylic acid, 2,2-dimethyl-3-2-methyl-1-propenyl-, ethyl ester, ethylchrysanthemate, ccris 2498, chrysanthemic acid, ethyl ester, ethyl 2,2-dimethyl-3-2-methyl-1-propenyl cyclopropanecarboxylate, ethyl 2,2-dimethyl-3-2-methylprop-1-en-1-yl cyclopropanecarboxylate, ethyl 2,2-dimethyl-3-2-methyl-1-propenyl cyclopropane-1-carboxylate
PubChem CID 7334
IUPAC Name ethyl 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate
SMILES CCOC(=O)C1C(C1(C)C)C=C(C)C

Specifications

Color Colorless to Yellow
Density 0.9000g/mL
Boiling Point 112°C (10.0 mmHg)
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4590 to 1.461
Quantity 25 g
Beilstein 09, II, 45
Specific Gravity 0.9
Formula Weight 196.29
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl chrysanthemate, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.