missing translation for 'onlineSavingsMsg'
Learn More

Ethyl chloroformate, 97%

Product Code. 11456730
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456730 5 g 5g
11466730 100 g 100g
11476730 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11456730 Supplier Thermo Scientific Alfa Aesar Supplier No. L06311.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 541-41-3 | C3H5ClO2 | 108.521 g/mol

Ethyl chloroformate is used as derivatization reagent to investigate stereochemical conversion of chiral non-steroidal anti-inflammatory drugs during derivatization reaction. It is also used with ammonia and cyanuric chloride to convert carboxylic acids to nitriles. It is used to develop flexible template strategy for the generation of nanometer-scale templates via dip-pen nanolithography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl chloroformate is used as derivatization reagent to investigate stereochemical conversion of chiral non-steroidal anti-inflammatory drugs during derivatization reaction. It is also used with ammonia and cyanuric chloride to convert carboxylic acids to nitriles. It is used to develop flexible template strategy for the generation of nanometer-scale templates via dip-pen nanolithography.

Solubility
Not miscible in water.

Notes
Protect from heat. Store away from moisture and oxidizing agents. Store under dry inert gas. Refrigerate. Incompatible with heat, moisture and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-41-3
Molecular Formula C3H5ClO2
Molecular Weight (g/mol) 108.521
MDL Number MFCD00000644
InChI Key RIFGWPKJUGCATF-UHFFFAOYSA-N
Synonym ethyl chloroformate, ethyl chlorocarbonate, cathyl chloride, ethoxycarbonyl chloride, chloroformic acid ethyl ester, ethylchloroformate, carbonochloridic acid, ethyl ester, chloro ethoxy methanone, ethylchloorformiaat, etil cloroformiato
PubChem CID 10928
IUPAC Name ethyl carbonochloridate
SMILES CCOC(=O)Cl

Specifications

Melting Point -81°C
Density 1.136
Boiling Point 92°C to 93°C
Flash Point 10°C (50°F)
Odor Pungent
Linear Formula ClCO2CH2CH3
Refractive Index 1.395
Quantity 5 g
UN Number UN1182
Beilstein 385653
Sensitivity Moisture sensitive
Merck Index 14,3784
Solubility Information Not miscible in water.
Formula Weight 108.52
Percent Purity 97%
Chemical Name or Material Ethyl chloroformate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.