missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Chloroacetate, TRC

Product Code. 30138691
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138691 1 g 1g
30135031 100 g 100g
30004798 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138691 Supplier Toronto Research Chemicals Supplier No. TRCE9017801G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-39-5
Molecular Formula C4 H7 Cl O2
Molecular Weight (g/mol) 122.55
Synonym Chloroacetic Acid-Ethyl Ester, Acetic acid, chloro-, ethyl ester (6CI,8CI,9CI), (Ethoxycarbonyl)methyl chloride, 2-Chloroacetic acid ethyl ester, Chloroacetic acid ethyl ester, Ethyl 2-chloroacetate, Ethyl 2-monochloroacetate, Ethyl chloracetate, Ethyl chloroacetate, Ethyl chloroethanoate, Ethyl monochloracetate, Ethyl monochloroacetate, Ethyl α-chloroacetate, NSC 8833
IUPAC Name ethyl 2-chloroacetate
SMILES CCOC(=O)CCl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H7ClO2/c1-2-7-4(6)3-5/h2-3H2,1H3
Formula Weight 122.0135
Chemical Name or Material Ethyl Chloroacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.