missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethyl cellulose, ethoxyl content 48%, 10 cps

Product Code. 10502411 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
500 g
Packaging:
Glass bottle
Unit Size:
250g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10502411 5 g Glass bottle 5g
10440471 250 g Glass bottle 250g
10450471 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10502411 Supplier Thermo Scientific Chemicals Supplier No. 190960050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 9004-57-3 | C23H24N6O4 | 448.483 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-57-3
Molecular Formula C23H24N6O4
Molecular Weight (g/mol) 448.483
MDL Number MFCD00131037
InChI Key ARSKJXYLLONUAJ-UHFFFAOYSA-N
Synonym n-butyl-n-2-acetoxyethyl-4-4-nitro-2,6-dicyanophenyl azo-3-methylbenzeneamine, 1,3-benzenedicarbonitrile, 2-4-2-acetyloxy ethyl butylamino-2-methylphenyl azo-5-nitro, 1,3-benzenedicarbonitrile, 2-2-4-2-acetyloxy ethyl butylamino-2-methylphenyl diazenyl-5-nitro, azo dye n5, 4-2,6-dicyano-4-nitrophenyl azo-3-methylphenyl amino hexyl acetate, ethyl cellulose, viscosity ethoxyl, 2-n-butyl-4-2,6-dicyano-4-nitrophenyl diazenyl-3-methylanilino ethyl acetate, 4-n-2-acetoxyethyl butyl amino-2',6'-dicyano-2-methyl-4'-nitroazobenzene
PubChem CID 166304
IUPAC Name 2-[N-butyl-4-[(2,6-dicyano-4-nitrophenyl)diazenyl]-3-methylanilino]ethyl acetate
SMILES CCCCN(CCOC(=O)C)C1=CC(=C(C=C1)N=NC2=C(C=C(C=C2C#N)[N+](=O)[O-])C#N)C

Specifications

Color White to Yellow
Physical Form Granules or Crystalline Powder
Merck Index 15, 3836
Quantity 5 g
Percent Purity 48 to 49.5% (ethoxyl)
Infrared Spectrum Authentic
Packaging Glass bottle
Viscosity 8 to 11cPs (5% soln. in 80/20 toluene/ethanol, 25°C)
Chemical Name or Material Ethyl cellulose, Ethoxyl Content 48%, 10 Cps

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.