missing translation for 'onlineSavingsMsg'
Learn More

Ethyl carbamate, 98%, Thermo Scientific Chemicals

Catalog Number 11430091
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
51-79-6
C3H7NO2
89.094
MFCD00007966
JOYRKODLDBILNP-UHFFFAOYSA-N
urethane, urethan, ethylurethane, carbamic acid ethyl ester, ethyl urethane, carbamic acid, ethyl ester, ethylcarbamate, pracarbamine, leucethane, pracarbamin
5641
CHEBI:17967
ethyl carbamate
CCOC(=O)N
This item is not returnable. View return policy
51-79-6
C3H7NO2
89.094
MFCD00007966
JOYRKODLDBILNP-UHFFFAOYSA-N
urethane, urethan, ethylurethane, carbamic acid ethyl ester, ethyl urethane, carbamic acid, ethyl ester, ethylcarbamate, pracarbamine, leucethane, pracarbamin
5641
CHEBI:17967
ethyl carbamate
CCOC(=O)N

Ethyl carbamate in its molten state is good solvent for various organic materials, preparation & modification of amino resins, as solubilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 48°C to 50°C
Color White
Boiling Point 182°C to 184°C
Flash Point >92°C (198°F)
Linear Formula H2NCO2C2H5
Quantity 50 g
Beilstein 635810
Merck Index 14,9877
Solubility Information Soluble in chloroform.
Formula Weight 89.09
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Ethyl carbamate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'