missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Caproate, TRC

Product Code. 30012457
Change view
Click to view available options
Quantity:
100 mg
50 g
Unit Size:
100mg
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012457 50 g 50g
17930895 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012457 Supplier Toronto Research Chemicals Supplier No. TRCE90160050G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-66-0
Molecular Formula C8 H16 O2
Molecular Weight (g/mol) 144.21
Synonym Hexanoic acid-ethyl ester, Hexanoic Acid Ethyl Ester, Hexanoic acid, ethyl ester (6CI, 8CI, 9CI, ACI), Caproic acid ethyl ester, Ethyl caproate, Ethyl capronate, Ethyl hexanoate, NSC 8882
IUPAC Name ethyl hexanoate
SMILES CCCCCC(=O)OCC

Specifications

Quantity 50 g
InChI Formula InChI=1S/C8H16O2/c1-3-5-6-7-8(9)10-4-2/h3-7H2,1-2H3
Formula Weight 144.115
Chemical Name or Material Caproic acid ethyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.