missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Caproate-d11, TRC

Product Code. 30387290
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387290 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30387290 Supplier Toronto Research Chemicals Supplier No. TRCE9016021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Caproate-d11
CAS 2159-19-5
Molecular Formula C8 2H11 H5 O2
Quantity 1 mg
Synonym Hexanoic-d11 acid, ethyl ester (7CI, 8CI, ACI), Ethyl hexanoate-2,2,3,3,4,4,5,5,6,6,6-d11, Hexanoic-2,2,3,3,4,4,5,5,6,6,6-d11 acid ethyl ester, Hexanoic-d11 acid ethyl ester, Ethyl hexanoate-d11, Hexanoic Acid Ethyl Ester-D11 (hexanoic-D11), Ethyl Caproate-d11
InChI Formula InChI=1S/C8H16O2/c1-3-5-6-7-8(9)10-4-2/h3-7H2,1-2H3/i1D3,3D2,5D2,6D2,7D2
SMILES [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)OCC
IUPAC Name ethyl 2,2,3,3,4,4,5,5,6,6,6-undecadeuteriohexanoate
Molecular Weight (g/mol) 155.28
Formula Weight 155.1841
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.