missing translation for 'onlineSavingsMsg'
Learn More

Ethyl caproate, 99+%

Product Code. 10731925 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10731925 100 mL Glass bottle 100mL
10081990 5 mL Glass bottle 5mL
10561801 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10731925 Supplier Thermo Scientific Acros Supplier No. 164991000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-66-0 | C8H16O2 | 144.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-66-0
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00009511
InChI Key SHZIWNPUGXLXDT-UHFFFAOYSA-N
Synonym ethyl caproate, ethyl hexoate, hexanoic acid, ethyl ester, hexanoic acid ethyl ester, capronic ether absolute, caproic acid ethyl ester, ethyl n-hexanoate, ethyl butyl acetate, caproate, acetic acid, butyl-, ethyl ester
PubChem CID 31265
ChEBI CHEBI:86055
IUPAC Name ethyl hexanoate
SMILES CCCCCC(=O)OCC

Specifications

Acidity 1mg KOH/g max.
Melting Point 67°C
Color Colorless
Density 0.8690g/mL
Boiling Point 168°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4CO2C2H5
Refractive Index 1.4060 to 1.409
Quantity 100 mL
Beilstein 02, 323
Merck Index 15, 3832
Specific Gravity 0.869
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in glycerol,soluble in alcohol and ether
Formula Weight 144.21
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Ethyl caproate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl caproate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.