missing translation for 'onlineSavingsMsg'
Learn More

Ethyl butyrylacetate, 98%

Product Code. 10061070 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10061070 25 g Glass bottle 25g
10438070 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10061070 Supplier Thermo Scientific Acros Supplier No. 118190250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3249-68-1 | C8H14O3 | 158.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3249-68-1
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.20
MDL Number MFCD00009401
InChI Key KQWWVLVLVYYYDT-UHFFFAOYSA-N
Synonym ethyl butyrylacetate, ethyl butyroacetate, hexanoic acid, 3-oxo-, ethyl ester, ethyl beta-ketohexanoate, ethyl alpha-butyrylacetate, 3-ketohexanoic acid ethyl ester, ethyl-3-oxohexanoate, unii-8q1ahg710e, ethyl .alpha.-butyrylacetate, fema no. 3683
PubChem CID 238498
ChEBI CHEBI:18119
IUPAC Name ethyl 3-oxohexanoate
SMILES CCCC(=O)CC(=O)OCC

Specifications

Color Colorless
Density 1.0000g/mL
Boiling Point 104.0°C (22.0 mmHg)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2COCH2CO2C2H5
Refractive Index 1.4265 to 1.4285
Quantity 25 g
Beilstein 03, 684
Specific Gravity 1
Solubility Information Solubility in water: immiscible with water
Formula Weight 158.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl butyrylacetate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.