missing translation for 'onlineSavingsMsg'
Learn More

Ethyl butyrate, 99%

Product Code. 11342336
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342336 1000 mL 1 L
11332336 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342336 Supplier Thermo Scientific Alfa Aesar Supplier No. L06025.AU
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 105-54-4 | C6H12O2 | 116.16 g/mol

Ethyl butyrate is used in flavors and fragrances. It is used as a solvent in perfumery products, and as a plasticizer for cellulose. It is used in the preparation of novel 2-cyanopyrimidines as cathespin K inhibitors. It is also used in the synthesis of pyridobenzimidazole derivatives exhibiting antifungal activity by the inhibition of β-1,6-glucan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl butyrate is used in flavors and fragrances. It is used as a solvent in perfumery products, and as a plasticizer for cellulose. It is used in the preparation of novel 2-cyanopyrimidines as cathespin K inhibitors. It is also used in the synthesis of pyridobenzimidazole derivatives exhibiting antifungal activity by the inhibition of β-1,6-glucan.

Solubility
Soluble in propylene glycol, paraffin oil, and kerosene.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-54-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009394
InChI Key OBNCKNCVKJNDBV-UHFFFAOYSA-N
Synonym ethyl butyrate, butyric acid ethyl ester, ethyl n-butyrate, butanoic acid, ethyl ester, butyric ester, butyric ether, butyric acid, ethyl ester, ethyl n-butanoate, butanoic acid ethyl ester, fema number 2427
PubChem CID 7762
IUPAC Name ethyl butanoate
SMILES CCCC(=O)OCC

Specifications

Melting Point -93°C
Density 0.879
Boiling Point 119°C to 120°C
Flash Point 26°C (79°F)
Refractive Index 1.392
Quantity 1000 mL
UN Number UN1180
Beilstein 506331
Merck Index 14,3775
Solubility Information Soluble in propylene glycol,paraffin oil,and kerosene.
Formula Weight 116.16
Percent Purity 99%
Chemical Name or Material Ethyl butyrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.