missing translation for 'onlineSavingsMsg'
Learn More

Ethyl bromoacetate, 98%

Product Code. 11468663
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468663 2500 g 2500g
11448663 100 g 100g
11458663 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468663 Supplier Thermo Scientific Alfa Aesar Supplier No. A10448.0E
  • ONLINE EXCLUSIVE PRICE
    £287.64
    £235.65 / 2500g
    Save £51.99
    18% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 105-36-2 | C4H7BrO2 | 167.002 g/mol

Ethyl bromoacetate is widely used as an alkylating reagent involved in the Reformatsky reaction to prepare the beta-hydroxy esters by reacting with carbonyl compounds. It is also used for the syntheses of witting reagent, artificial diethylstilbestrol antigen, 3-phenyl-1-naphthol and steroidal thiazolidinone derivatives. It finds applications in the preparation of reversibly photoresponsive coumarin-stabilized polymeric nanoparticles in an aqueous medium which act as a detectable drug carrier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl bromoacetate is widely used as an alkylating reagent involved in the Reformatsky reaction to prepare the beta-hydroxy esters by reacting with carbonyl compounds. It is also used for the syntheses of witting reagent, artificial diethylstilbestrol antigen, 3-phenyl-1-naphthol and steroidal thiazolidinone derivatives. It finds applications in the preparation of reversibly photoresponsive coumarin-stabilized polymeric nanoparticles in an aqueous medium which act as a detectable drug carrier.

Solubility
Miscible with ethanol, acetone, benzene and ethyl ether. Immiscible with water.

Notes
It is a lachrymator. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-36-2
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.002
MDL Number MFCD00000191
InChI Key PQJJJMRNHATNKG-UHFFFAOYSA-N
Synonym ethyl bromoacetate, antol, ethyl bromacetate, bromoacetic acid ethyl ester, ethyl monobromoacetate, acetic acid, bromo-, ethyl ester, ethoxycarbonylmethyl bromide, bromoacetic acid, ethyl ester, ethyl alpha-bromoacetate, ethyl bromo acetate
PubChem CID 7748
IUPAC Name ethyl 2-bromoacetate
SMILES CCOC(=O)CBr

Specifications

Melting Point -38°C
Density 1.506
Boiling Point 159°C
Flash Point 47°C (117°F)
Assay Percent Range 98%
Linear Formula BrCH2CO2CH2CH3
Refractive Index 1.451
Quantity 2500 g
UN Number UN1603
Solubility Information Miscible with ethanol,acetone,benzene and ethyl ether. Immiscible with water.
Formula Weight 167
Percent Purity 98%
Chemical Name or Material Ethyl bromoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.