missing translation for 'onlineSavingsMsg'
Learn More

Ethyl bromoacetate, 98%

Product Code. 10429640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10429640 100 g Glass bottle 100g
10439640 500 g Glass bottle 500g
10175633 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10429640 Supplier Thermo Scientific Acros Supplier No. 158591000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 105-36-2 | C4H7BrO2 | 167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-36-2
MDL Number MFCD00000191
InChI Key PQJJJMRNHATNKG-UHFFFAOYSA-N
Synonym ethyl bromoacetate, antol, ethyl bromacetate, bromoacetic acid ethyl ester, ethyl monobromoacetate, acetic acid, bromo-, ethyl ester, ethoxycarbonylmethyl bromide, bromoacetic acid, ethyl ester, ethyl alpha-bromoacetate, ethyl bromo acetate
PubChem CID 7748
IUPAC Name ethyl 2-bromoacetate
SMILES CCOC(=O)CBr

Specifications

Melting Point -38.0°C
Density 1.5060g/mL
Boiling Point 159.0°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C4H7BrO2
Linear Formula BrCH2CO2C2H5
Refractive Index 1.4500 to 1.4520
Quantity 100 g
Beilstein 02, 214
Fieser 02,192; 13,60
Specific Gravity 1.506
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol, benzene and ether
Molecular Weight (g/mol) 167
Formula Weight 167
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl bromoacetate, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl bromoacetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H310-Fatal in contact with skin.
  • H300-Fatal if swallowed.
  • H330-Fatal if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.