missing translation for 'onlineSavingsMsg'
Learn More

Ethyl bromoacetate, 98%, Thermo Scientific Chemicals

Catalog Number 11448663
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
2500 g
105-36-2
C4H7BrO2
167.002
MFCD00000191
PQJJJMRNHATNKG-UHFFFAOYSA-N
ethyl bromoacetate, antol, ethyl bromacetate, bromoacetic acid ethyl ester, ethyl monobromoacetate, acetic acid, bromo-, ethyl ester, ethoxycarbonylmethyl bromide, bromoacetic acid, ethyl ester, ethyl alpha-bromoacetate, ethyl bromo acetate
7748
ethyl 2-bromoacetate
CCOC(=O)CBr
105-36-2
C4H7BrO2
167.002
MFCD00000191
PQJJJMRNHATNKG-UHFFFAOYSA-N
ethyl bromoacetate, antol, ethyl bromacetate, bromoacetic acid ethyl ester, ethyl monobromoacetate, acetic acid, bromo-, ethyl ester, ethoxycarbonylmethyl bromide, bromoacetic acid, ethyl ester, ethyl alpha-bromoacetate, ethyl bromo acetate
7748
ethyl 2-bromoacetate
CCOC(=O)CBr

Ethyl bromoacetate is widely used as an alkylating reagent involved in the Reformatsky reaction to prepare the beta-hydroxy esters by reacting with carbonyl compounds. It is also used for the syntheses of witting reagent, artificial diethylstilbestrol antigen, 3-phenyl-1-naphthol and steroidal thiazolidinone derivatives. It finds applications in the preparation of reversibly photoresponsive coumarin-stabilized polymeric nanoparticles in an aqueous medium which act as a detectable drug carrier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl bromoacetate is widely used as an alkylating reagent involved in the Reformatsky reaction to prepare the beta-hydroxy esters by reacting with carbonyl compounds. It is also used for the syntheses of witting reagent, artificial diethylstilbestrol antigen, 3-phenyl-1-naphthol and steroidal thiazolidinone derivatives. It finds applications in the preparation of reversibly photoresponsive coumarin-stabilized polymeric nanoparticles in an aqueous medium which act as a detectable drug carrier.

Solubility
Miscible with ethanol, acetone, benzene and ethyl ether. Immiscible with water.

Notes
It is a lachrymator. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point °C to 38°C
Density 1.506 g/mL
Boiling Point 159°C
Flash Point 47°C (117°F)
Assay Percent Range 98%
Linear Formula BrCH2CO2CH2CH3
Refractive Index 1.451
Quantity 100 g
UN Number UN1603
Solubility Information Miscible with ethanol,acetone,benzene and ethyl ether. Immiscible with water.
Formula Weight 167
Percent Purity 98%
Chemical Name or Material Ethyl bromoacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'