missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Biotin, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl Biotin |
| Analyte or Component Names | Ethyl Biotin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H20 N2 O3 S |
| Synonym | 2-Ethyl-5-[(3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanoic Acid |
| InChI Formula | InChI=1S/C12H20N2O3S/c1-2-7(11(15)16)4-3-5-9-10-8(6-18-9)13-12(17)14-10/h7-10H,2-6H2,1H3,(H,15,16)(H2,13,14,17)/t7?,8-,9-,10-/m0/s1 |
| Solution Type | Neat |
| SMILES | CCC(CCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12)C(=O)O |
| IUPAC Name | 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-2-ethyl-pentanoic acid |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?